missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF701 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552158
This item is not returnable.
View return policy
Description
Immunogen sequence: QIHASHHIGD TCFQEIEKDI HDFVFQWQEN ETNGHEALMT KTKKLMSSTE RHDQRHAGNK PIKNELGSSF HSHLPEVHIF HPEGKIGNQV EKAINDAFSV SASQRISCRP KTRISNKYRN NFLQSSLLTQ Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 26%.
ZNF701 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 7 C2H2-type zinc fingers and KRAB domain. ZNF701 may be involved in transcriptional regulation.
Specifications
| ZNF701 | |
| Polyclonal | |
| Unconjugated | |
| ZNF701 | |
| Zinc finger protein 701; ZNF701 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NV72 | |
| ZNF701 | |
| Recombinant protein corresponding to Human ZNF701. Recombinant protein control fragment (Product #RP-99727). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction