missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF700 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ZNF700 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF700 Polyclonal specifically detects ZNF700 in Human samples. It is validated for Western Blot.Specifications
| ZNF700 | |
| Polyclonal | |
| Rabbit | |
| NP_653167 | |
| 90592 | |
| Synthetic peptide directed towards the C terminal of human ZNF700The immunogen for this antibody is ZNF700. Peptide sequence CKECEKAFCKFSSFQIHERKHRGEKPYECKHCGNGFTSAKILQIHARTHI. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| zinc finger protein 700 | |
| ZNF700 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title