missing translation for 'onlineSavingsMsg'
Learn More

ZNF70 Antibody (1D8), Novus Biologicals™

Product Code. 18400280 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400280 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400280 Supplier Novus Biologicals Supplier No. H00007621M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF70 Monoclonal antibody specifically detects ZNF70 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF70
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1D8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH40161
Gene Alias Cos17, MGC48959, zinc finger protein 70, zinc finger protein 70 (Cos17), Zinc finger protein N27C7-1
Host Species Mouse
Immunogen ZNF70 (AAH40161, 1 a.a. ~ 446 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEVPPATKFGETFAFENRLESQQGLFPGEDLGDPFLQERGLEQMAVIYKEIPLGEQDEENDDYEGNFSLCSSPVQHQSIPPGTRPQDDELFGQTFLQKSDLSMCQIIHSEEPSPCDCAETDRGDSGPNAPHRTPQPAKPYACRECGKAFSQSSHLLRHLVIHTGEKPYECCECGKAFSQSSHLLRHQIIHTGEKPYECRECGKAFRQSSALTQHQKIHTGKRPYECRECGKDFSRSSSLRKHERIHTGERPYQCKECGKSFNQSSGLSQHRKIHTLKKPHECDLCGKAFCHRSHLIRHQRIHTGKKPYKCDECGKAFSQSSNLIEHRKTHTGEKPYKCQKCGKAFSQSSSLIEHQRIHTGEKPYECCQCGKAFCHSSALIQHQRIHTGKKPYTCECGKAFRHRSALIEHYKTHTREKPYVCNLCGKSFRGSSHLIRHQKIHSGEKL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7621
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.