missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521521
This item is not returnable.
View return policy
Description
Recommended positive controls: A431. Predicted reactivity: Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in transcriptional regulation.
Specifications
| ZNF7 | |
| Polyclonal | |
| Unconjugated | |
| ZNF7 | |
| C2-H2 type zinc finger protein; HF.16; KOX4; zf30; zinc finger protein 7; zinc finger protein 7 (KOX 4, clone HF.16); zinc finger protein HF.16; Zinc finger protein KOX4; zinc finger protein zfp30; Zinc finger protein-7 (KOX4); ZNF7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P17097 | |
| ZNF7 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction