missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF690 Antibody (ZSCAN29/2610), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79881PCP
This item is not returnable.
View return policy
Description
ZNF690 Monoclonal antibody specifically detects ZNF690 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| ZNF690 | |
| Monoclonal | |
| PerCP | |
| EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690 | |
| Recombinant full-length human ZNF690 protein (Uniprot: Q8IWY8) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| ZSCAN29/2610 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 146050 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction