missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF690 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78200
This item is not returnable.
View return policy
Description
ZNF690 Polyclonal specifically detects ZNF690 in Human samples. It is validated for Immunoprecipitation.
Specifications
| ZNF690 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 146050 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-10 mcg/mg | |
| NP_689668.3 | |
| ZSCAN29 | |
| The immunogen for this product maps to a region between residue 600 and 650 of human Zinc Finger and SCAN Domain Containing 29 using the numbering given in entry NP_689668.3 (GeneID 146050). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction