missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF690 Polyclonal antibody specifically detects ZNF690 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF690 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 802 and 852 of human Zinc Finger and SCAN Domain Containing 29 using the numbering given in entry NP_689668.3 (GeneID 146050). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?