missing translation for 'onlineSavingsMsg'
Learn More

ZNF69 Antibody (OTI3C3) - Azide and BSA Free, Novus Biologicals™

Product Code. 18391251 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391251 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391251 Supplier Novus Biologicals Supplier No. NBP274954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF69 Monoclonal specifically detects ZNF69 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF69
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias Cos5, hZNF3, MGC59928, zinc finger protein 69, zinc finger protein 69 (Cos5)
Gene Symbols ZNF69
Host Species Mouse
Immunogen Full length human recombinant protein of human ZNF69(NP_068734) produced in E.coli.
Molecular Weight of Antigen 17.2 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7620
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.