missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF69 Antibody (OTI3C3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74954AF594
This item is not returnable.
View return policy
Description
ZNF69 Monoclonal antibody specifically detects ZNF69 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZNF69 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Cos5, hZNF3, MGC59928, zinc finger protein 69, zinc finger protein 69 (Cos5) | |
| Full length recombinant protein of human ZNF69 (NP_068734) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3C3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 7620 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction