missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF677 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ZNF677 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF677 Polyclonal specifically detects ZNF677 in Human samples. It is validated for Western Blot.Specifications
| ZNF677 | |
| Polyclonal | |
| Rabbit | |
| Q86XU0 | |
| 342926 | |
| Synthetic peptides corresponding to ZNF677 (zinc finger protein 677) The peptide sequence was selected from the N terminal of ZNF677. Peptide sequence YRNLLSLDEDNIPPEDDISVGFTSKGLSPKENNKEELYHLVILERKESHG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MGC48625, zinc finger protein 677 | |
| ZNF677 | |
| IgG | |
| 68 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title