missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF670 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541193
This item is not returnable.
View return policy
Description
Peptide sequence: EDQNIQDDFK NPGRNLSSHV VERLFEIKEG SQYGETFSQD SNLNLNKKVS Sequence homology: Human: 100%.
This gene, ZNF670, which is located on chromosome 1, is predicted to encode a zinc finger protein, currently with unknown fucntion.
Specifications
| ZNF670 | |
| Polyclonal | |
| Unconjugated | |
| ZNF670 | |
| Zinc finger protein 670; ZNF670 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 93474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BS34 | |
| ZNF670 | |
| Synthetic peptide directed towards the N-terminal of human ZNF670 (aa 48-97). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction