missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF670 Monoclonal Antibody (OTI3E1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527403
This item is not returnable.
View return policy
Description
ZNF670 Monoclonal Antibody for Western Blot, IHC (P)
This gene, ZNF670, which is located on chromosome 1, is predicted to encode a zinc finger protein, currently with unknown fucntion.
Specifications
| ZNF670 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BS34 | |
| ZNF670 | |
| Full length human recombinant protein of ZNF670 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E1 | |
| Unconjugated | |
| ZNF670 | |
| Zinc finger protein 670; ZNF670 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 93474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction