missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF667 Polyclonal antibody specifically detects ZNF667 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ZNF667 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686O111, FLJ14011, FLJ45518, MIPU1, myocardial ischemic preconditioning upregulated 1 ortholog, zinc finger protein 667 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human MIPU1. The immunogen is located within the first 50 amino acids of MIPU1. Amino Acid Squence: SQEEWEWLSPIQKDLYE |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?