missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF654 Polyclonal antibody specifically detects ZNF654 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF654 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ10997, FLJ21142, zinc finger protein 654 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 477 to 527 of human Zinc Finger Protein 654 using the numbering given in entry NP_060763.2 (GeneID 55279). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?