missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF645 Monoclonal Antibody (K92039_7G8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556051
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: FTRKHGNLTV DHIQNNSDSG AKKPTPPDYY PECQSQPAVS SPHHIIPQKQ HYAPPPSPSS PVNHQMPYPP QDVVTPNSVR SQVPALTTTY DPSSGYIIVK VPPDMNSPPL RAPQSQNGNP
ZNF645 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions.
Specifications
| ZNF645 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8N7E2 | |
| ZNF645 | |
| Recombinant human CBLL2 (immunogen range is 231-350). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K92039_7G8 | |
| Unconjugated | |
| ZNF645 | |
| Cbl proto-oncogene-like protein 2; CBLL2; c-Cbl-like protein 2; CT138; E3 ubiquitin ligase; E3 ubiquitin-protein ligase CBLL2; E3 ubiquitin-protein ligase ZNF645; HAKAIL; RING-type E3 ubiquitin transferase ZNF645; Zinc finger protein 645; ZNF645 | |
| Mouse | |
| Protein A | |
| RUO | |
| 158506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction