missing translation for 'onlineSavingsMsg'
Learn More

ZNF644 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18407386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407386 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407386 Supplier Novus Biologicals Supplier No. NBP178195FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF644 Polyclonal antibody specifically detects ZNF644 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF644
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BM-005, KIAA1221Zep-2, MGC60165, MGC70410, ZEP2, zinc finger motif enhancer binding protein 2, Zinc finger motif enhancer-binding protein 2, zinc finger protein 644
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Zinc Finger Protein 644 using the numbering given in entry NP_958357.1 (GeneID 84146).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84146
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.