missing translation for 'onlineSavingsMsg'
Learn More

ZNF638 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£155.00 - £364.00

Specifications

Antigen ZNF638
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18691862
View Documents
Novus Biologicals
NBP2-94191-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18696742
View Documents
Novus Biologicals
NBP2-94191-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

ZNF638 Polyclonal antibody specifically detects ZNF638 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

ZNF638
Western Blot
Unconjugated
Rabbit
Human, Mouse
CTCL tumor antigen se33-1, CTCL-associated antigen se33-1, Cutaneous T-cell lymphoma-associated antigen se33-1, DKFZp686P1231, MGC26130, MGC90196, NP220 nuclear protein, NP220zinc finger, matrin-like, Nuclear protein 220, ZFML, Zfp638, Zinc finger matrin-like protein, zinc finger protein 638
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZNF638 (NP_001239542.1). MSRPRFNPRGDFPLQRPRAPNPSGMRPPGPFMRPGSMGLPRFYPAGRARGIPHRFAGHESYQNMGPQRMNVQVTQHRTDPRLTKEKLDFHEAQQKKGKPHGSRWDDEPHISASVAVKQSSVTQVTEQSPKVQSRYTKESASSILASFGLSNEDLEELSRYPDEQLTPENMPLILRDIRMRKMGRRLPNLPSQSRNKETLGSEAVSSNVIDYGHASKYGYTEDPLEVRIYDPEIPTDEVENEFQSQQNISASVPNPNVICNSMFPVEDVFRQMDFPGESSNNRSFFSVESGTKMSGLHISG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
27332
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.