missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF622 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557698
This item is not returnable.
View return policy
Description
Immunogen sequence: VAHMTKDHSF FIPDIEYLSD IKGLIKYLGE KVGVGKICLW CNEKGKSFYS TEAVQAHMND KSHCKLFTDG DAALEFADFY DFRSSYPDHK EGE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.
Specifications
| ZNF622 | |
| Polyclonal | |
| Unconjugated | |
| ZNF622 | |
| 1110033B05Rik; D15Ertd806e; liver regeneration-related protein LRRG121; Zfp622; Zinc finger protein 622; zinc finger-like protein 9; Znf622; ZPR9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 90441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969S3 | |
| ZNF622 | |
| Recombinant protein corresponding to Human ZNF622. Recombinant protein control fragment (Product #RP-102723). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction