missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF622 Antibody (PCRP-ZNF622-1C11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08811FR
This item is not returnable.
View return policy
Description
ZNF622 Monoclonal antibody specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| ZNF622 | |
| Monoclonal | |
| DyLight 680 | |
| MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
| Recombinant full-length human ZNF622 protein (Uniprot: Q969S3) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-ZNF622-1C11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 90441 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction