missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF622 Polyclonal antibody specifically detects ZNF622 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF622 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human Zinc Finger Protein 622 using the numbering given in entry NP_219482.1 (GeneID 90441). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?