missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF610 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563175

Product Code. 15750952

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: YIRGKSYEYE CSEDGEVFRV PASLTNHQ Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF610
Polyclonal
Unconjugated
ZNF610
FLJ36040; Zinc finger protein 610; zink finger protein; ZNF610
Rabbit
Antigen affinity chromatography
RUO
162963
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N9Z0
ZNF610
Recombinant protein corresponding to Human ZNF610. Recombinant protein control fragment (Product #RP-101794).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.