missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF610 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585051
This item is not returnable.
View return policy
Description
Immunogen sequence: KVFNQNPHLS RHRKIHAGEN SLRTLQME Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 46%.
May be involved in transcriptional regulation.
Specifications
| ZNF610 | |
| Polyclonal | |
| Unconjugated | |
| ZNF610 | |
| FLJ36040; Zinc finger protein 610; zink finger protein; ZNF610 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 162963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N9Z0 | |
| ZNF610 | |
| Recombinant protein corresponding to Human ZNF610. Recombinant protein control fragment (Product #RP-108700). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction