missing translation for 'onlineSavingsMsg'
Learn More

ZNF607 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen ZNF607
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18278597
View Documents
Novus Biologicals
NBP1-80125
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF607 Polyclonal specifically detects ZNF607 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF607
Polyclonal
Rabbit
Human
FLJ14802, MGC13071, zinc finger protein 607
ZNF607
IgG
81 kDa
Western Blot
Unconjugated
RUO
NP_116078
84775
Synthetic peptide directed towards the middle region of human ZNF607. Peptide sequence KECGKGFTCRYQLTMHQRIYSGEKHYECKENGEAFSSGHQLTAPHTFESV.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.