missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF606 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54926
This item is not returnable.
View return policy
Description
ZNF606 Polyclonal specifically detects ZNF606 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF606 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| KIAA1852FLJ14260, Zinc finger protein 328, zinc finger protein 606, ZNF328 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80095 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ZNF606 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:WVRNLESKALIPAQSIFEEEQSHGMKLERYIWDDPWFSRLEVLGCKDQLEMYHMNQSTAMRQMVFMQKQV | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction