missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF599 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589017
This item is not returnable.
View return policy
Description
Immunogen sequence: ETCRLLVSLG HPVPKPELIY LLEHGQELWT VKRGLSQSTC AGEKAKPKIT EPTASQLAFS EESSFQELLA QRSSRDSRLG QARDEEKLIK IQEGNLRPGT NPHKEICPEK LSYKHDDLEP DDSLGLRVLQ ERVTPQDALH ECDSQGPGKD PMTDARNNPY T; Positive Samples: 293T; Cellular Location: Nucleus.
May be involved in transcriptional regulation.
Specifications
| ZNF599 | |
| Polyclonal | |
| Unconjugated | |
| ZNF599 | |
| Zinc finger protein 599; ZNF599 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 148103 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96NL3 | |
| ZNF599 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-200 of human ZNF599 (NP_0010072491). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction