missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF598 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531409
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Zebrafish (85%), Xenopus laevis (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
Specifications
| ZNF598 | |
| Polyclonal | |
| Unconjugated | |
| ZNF598 | |
| BC023040; C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Ntrap; Zfp598; Zinc finger protein 598; ZNF598 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 213753, 90850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.29 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q80YR4, Q86UK7 | |
| Zfp598, ZNF598 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF598. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction