missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF598 Monoclonal Antibody (K92040_5C9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555719
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: PRHSRRNEGV VGGEDYEEVD RYSRQGRVAR AGTRGAQQSR RGSWRYKREE EDREVAAAVR ASVAAQQQEE ARRSEDQEEG GRPKKEEAAA RGPEDPRGPR RSPRTQGEGP GPKETSTNGP VSQEAFSVTG
ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
Specifications
| ZNF598 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q86UK7 | |
| ZNF598 | |
| Recombinant human ZNF598 (immunogen range is 291-420). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K92040_5C9 | |
| Unconjugated | |
| ZNF598 | |
| BC023040; C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Ntrap; Zfp598; Zinc finger protein 598; ZNF598 | |
| Mouse | |
| Protein A | |
| RUO | |
| 90850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction