missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF598 Monoclonal Antibody (1E2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522840
This item is not returnable.
View return policy
Description
Peptide Sequence: SCVLCCGDLE ATALGRCDHP VCYRCSTKMR VLCEQRYCAV CREELRQVVF GKKLPAFATI PIHQLQHEKK YDIYFADGKV YALYRQLLQH ECPRCPELP.
ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
Specifications
| ZNF598 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q86UK7 | |
| ZNF598 | |
| ZNF598 (NP_835461, 28 a.a. ~ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry | |
| 1E2 | |
| Unconjugated | |
| ZNF598 | |
| BC023040; C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Ntrap; Zfp598; Zinc finger protein 598; ZNF598 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 90850 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction