missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF596 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | ZNF596 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653859
|
Novus Biologicals
NBP2-49183-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665897
|
Novus Biologicals
NBP2-49183 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF596 Polyclonal antibody specifically detects ZNF596 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ZNF596 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ36123, zinc finger protein 596 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LVSIGKQLCKSVVLSQLEQVEKLSTQRISLLQGREVGIKHQEIPFIQHIYQKGTSTISTMRSHTQEDP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 169270 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title