missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF593 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565865
This item is not returnable.
View return policy
Description
Immunogen sequence: DSTNLKTHFRS KDHKKRLKQL SVEPYSQEEA ERAAGMGSYV PPRRLAVPTE VSTEVPEMDT ST Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
Specifications
| ZNF593 | |
| Polyclonal | |
| Unconjugated | |
| ZNF593 | |
| 3110024A21Rik; AV062409; E130106C14Rik; Zfp593; Zinc Finger 593; Zinc finger protein 593; zinc finger protein T86; ZNF593; ZT86 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51042 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00488 | |
| ZNF593 | |
| Recombinant Human ZNF593. Recombinant protein control fragment (Product #RP-106536). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction