Learn More
ZNF592 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554467
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LARELGTCSS VPPRQRLKPA HSKLSSCVAA LVALQAKRVA SVTKEDQPGH TKDLSGPTKE SSKGSPKMPK SPKSPRSPLE ATRKSIKPSD SPRSICSDSS SKGSPSVAAS S Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia. [provided by RefSeq, Jan 2011].Specifications
ZNF592 | |
Polyclonal | |
Unconjugated | |
ZNF592 | |
8430405N24; A730014M16Rik; CAMOS; KIAA0211; mKIAA0211; SCAR5; Zfp592; Zfp-592; Zinc finger protein 592; ZNF592 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92610 | |
ZNF592 | |
Recombinant protein corresponding to Human ZNF592 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only