missing translation for 'onlineSavingsMsg'
Learn More

ZNF592 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-74658IR

Product Code. 18722160

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF592 Polyclonal antibody specifically detects ZNF592 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF592
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin
CAMOS, KIAA0211MGC138437, MGC138439, SCAR5, spinocerebellar ataxia, autosomal recessive 5, zinc finger protein 592
The immunogen recognized by this antibody maps to a region between residue 950 and 1000 of human zinc finger protein 592 using the numbering given in entry NP_055445.2 (GeneID 9640).
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
9640
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.