missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF585B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ZNF585B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF585B Polyclonal specifically detects ZNF585B in Human samples. It is validated for Western Blot.Specifications
| ZNF585B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ14928, FLJ33255, SZFP41, zinc finger protein 41-like protein, zinc finger protein 585B | |
| ZNF585B | |
| IgG | |
| 88 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_689492 | |
| 92285 | |
| Synthetic peptide directed towards the N terminal of human ZNF585B. Peptide sequence RKIIGYKPASSQDQKIYSGEKSYECAEFGKSFTWKSQFKVHLKVPTGEKL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title