missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF579 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569047
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.
May be involved in transcriptional regulation.
Specifications
| ZNF579 | |
| Polyclonal | |
| Unconjugated | |
| ZNF579 | |
| 1110003A17Rik; FLJ35453; Zfp579; Zinc finger protein 579; ZNF579 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 163033 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NAF0 | |
| ZNF579 | |
| synthetic peptide directed towards the middle region of human ZNF579. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction