missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF572 Monoclonal Antibody (OTI2F6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527510
This item is not returnable.
View return policy
Description
ZNF572 Monoclonal Antibody for Western Blot
May be involved in transcriptional regulation.
Specifications
| ZNF572 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z3I7 | |
| ZNF572 | |
| Full length human recombinant protein of ZNF572 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2F6 | |
| Unconjugated | |
| ZNF572 | |
| Zinc finger protein 572; ZNF572 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 137209 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction