missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF569 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562713

Product Code. 15760502

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EIWGVDEHQK NQDRLLRQVE VKFQKTLTEE KGNECQKKFA NVFPLNSDFF PSRHNLYEYD LFGKCLEHNF DCHNNVKCLM RKEHCEYNEP VKSYGNSSSH FVIT Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 44%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF569
Polyclonal
Unconjugated
ZNF569
2810054M15Rik; DNA-binding protein; KRAB8; LOW QUALITY PROTEIN: zinc finger protein 569; Mszf21; mszf77; Unknown (protein for IMAGE:7986680); ZAP1; Zfp66; Zfp74; Zfp-74; zinc finger protein 568; zinc finger protein 569; Zinc finger protein 74; ZNF; Znf569
Rabbit
Antigen affinity chromatography
RUO
148266
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
0.8 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5MCW4
ZNF569
Recombinant protein corresponding to Human ZNF569. Recombinant protein control fragment (Product #RP-92441).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.