missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF563 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | ZNF563 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663880
|
Novus Biologicals
NBP2-93199-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694291
|
Novus Biologicals
NBP2-93199-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF563 Polyclonal antibody specifically detects ZNF563 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZNF563 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ34797, zinc finger protein 563 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ZNF563 (NP_660319.1). MDAVAFEDVAVNFTQEEWALLGPSQKNLYRYVMQETIRNLDCIRMIWEEQNTEDQYKNPRRNLRCHMVERFSESKDSSQCGETFSLIRDSIVNNSICPGE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 147837 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title