missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF562 Monoclonal antibody specifically detects ZNF562 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | ZNF562 |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-ZNF562-1A1 |
| Conjugate | DyLight 650 |
| Dilution | Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ20079, MGC126160, zinc finger protein 562 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human ZNF562 protein (Uniprot: Q6V9R5) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?