missing translation for 'onlineSavingsMsg'
Learn More

ZNF551 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-86513-0.1ML

Product Code. 18306521

  • £361.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF551 Polyclonal specifically detects ZNF551 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF551
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
90233
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
DKFZp686H1038, KOX 23 protein (56 AA), KOX23, MGC52307, zinc finger protein 551, zinc finger protein KOX23
The immunogen is a synthetic peptide directed towards the N-terminal region of Human ZNF551
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.