missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF543 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06246-100ul
This item is not returnable.
View return policy
Description
ZNF543 Polyclonal antibody specifically detects ZNF543 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF543 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DKFZp434H055, MGC119382, MGC119384, zinc finger protein 543 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF543 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 125919 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction