missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF521 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554898
This item is not returnable.
View return policy
Description
Immunogen sequence: VTVGYTSVSS TTPDSNLSVD SSTMVEAAPP IPKSRGRKRA AQQTPDMTGP SSKQAKVTYS CIYCNKQLFS SLAVLQIHLK TMH Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The zinc finger protein 521 (ZNF521) is a transcription factor containing an N-terminal transcriptional repressor motif and 30 zinc finger domains. It plays a role in both erythroid cell and osteoblast differentiation during development, inhibiting the activities of early B-cell factor 1 (EBF1) in erythroid cells and Runx2 in osteoblast precursors. ZFP521 binds to both Runx2 and histone deacetylase 3 (HDAC3), promotes their association and antagonizes Runx2 transcriptional activity in a HDAC3-dependent manner, thereby regulating osteoblast differentiation, skeletal development, and bone homeostasis.
Specifications
| ZNF521 | |
| Polyclonal | |
| Unconjugated | |
| ZNF521 | |
| B930086A16Rik; early hematopoietic zinc finger protein; Ecotropic viral integration site 3 protein; EHZF; Evi3; LIP3; LYST-interacting protein 3; Zfp521; Zinc finger protein 521; Znf521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96K83 | |
| ZNF521 | |
| Recombinant protein corresponding to Human ZNF521. Recombinant protein control fragment (Product #RP-93654). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction