missing translation for 'onlineSavingsMsg'
Learn More

ZNF521 Antibody, Allophycocyanin, Novus Biologicals™

Product Code. 18403342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403342 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403342 Supplier Novus Biologicals Supplier No. NBP191270APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF521 Polyclonal antibody specifically detects ZNF521 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF521
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation PBS
Gene Alias EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250) [UniProt Q96K83]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25925
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.