missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF512 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24508-1-AP-20UL
This item is not returnable.
View return policy
Description
ZNF512 may be involved in transcriptional regulation.Specifications
| ZNF512 | |
| Polyclonal | |
| Unconjugated | |
| ZNF512 | |
| KIAA1805, zinc finger protein 512, ZNF512 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269639, 84450 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69Z99, Q96ME7 | |
| ZNF512 | |
| ZNF512 Fusion Protein Ag21602 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction