missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF509 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21772-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
| ZNF509 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB49 | |
| ZBTB49, zinc finger protein 509, ZNF509 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 166793, 75079 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6ZSB9, Q8BXX2 | |
| Zbtb49 | |
| ZNF509 Fusion Protein Ag16445 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction