Learn More
ZNF509 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21772-1-AP-150UL
 View more versions of this product
Product Code. 16861365
  • £349.00 / 150µL
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF509
Polyclonal
Unconjugated
ZBTB49
ZBTB49, zinc finger protein 509, ZNF509
Rabbit
Antigen Affinity Chromatography
RUO
166793, 75079
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6ZSB9, Q8BXX2
Zbtb49
ZNF509 Fusion Protein Ag16445
150 μL
Primary
Human, Mouse