missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF507 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78194AF647
This item is not returnable.
View return policy
Description
ZNF507 Polyclonal antibody specifically detects ZNF507 in Human samples. It is validated for Western Blot
Specifications
| ZNF507 | |
| Polyclonal | |
| Western Blot | |
| BMZF-1ZNF411, Bone marrow zinc finger 1, DNA-binding protein, FLJ90391, zinc finger protein 253, Zinc finger protein 411BMZF1 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Zinc Finger Protein 507 using the numbering given in entry NP_055725.2 (GeneID 22847). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 22847 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction