missing translation for 'onlineSavingsMsg'
Learn More

ZNF503 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18428267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428267 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428267 Supplier Novus Biologicals Supplier No. NBP232130AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF503 Polyclonal antibody specifically detects ZNF503 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF503
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 150 to 200 of human Zinc Finger Protein 503 using the numbering given in entry NP_116161.2 (GeneID 84858).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84858
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.