missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF502 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555173
This item is not returnable.
View return policy
Description
Immunogen sequence: MLNMQGAEER DIRRETCPGW VNKNKPALEQ DVCKIDSSGI VVKRFQEDEY QDSTFEEKYA CEGMKENSPR EIAES Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 25%.
May be involved in transcriptional regulation.
Specifications
| ZNF502 | |
| Polyclonal | |
| Unconjugated | |
| ZNF502 | |
| zinc finger protein 502; ZNF502 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 91392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TBZ5 | |
| ZNF502 | |
| Recombinant protein corresponding to Human ZNF502. Recombinant protein control fragment (Product #RP-93640). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction