missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF502 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93235-0.1ml
This item is not returnable.
View return policy
Description
ZNF502 Polyclonal antibody specifically detects ZNF502 in Human samples. It is validated for Western Blot
Specifications
| ZNF502 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human ZNF502 (NP_149987.2). MLNMQGAEERDIRRETCPGWVNKNKPALEQDVCKIDSSGIVVKRFQEDEYQDSTFEEKYACEGMKENSPREIAESCLFQEGGFGRITFIHKEAPPEIISQGYNFEKSLLLTSSLVTRLRVSTEESLHQWETSNIQTNDISDQSKCPTLCT | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 91392 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction