missing translation for 'onlineSavingsMsg'
Learn More

ZNF500 Antibody (OTI2D2), DyLight 405, Novus Biologicals™

Product Code. 18343560 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343560 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343560 Supplier Novus Biologicals Supplier No. NBP274953V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF500 Monoclonal specifically detects ZNF500 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF500
Applications Western Blot
Classification Monoclonal
Clone OTI2D2
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238
Gene Symbols ZNF500
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 102-325 of human ZNF500 (NP_067678) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26048.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.